missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Mycoplasma / Ureaplasma Broth

Product Code. 10536203
Change view
Click to view available options
Description:
Mycoplasma / Ureaplasma Broth
Quantity:
10/Pk.
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Quantity unitSize
10536203 Mycoplasma / Ureaplasma Broth 10/Pk. Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10536203 Supplier Thermo Scientific™ Supplier No. TV5081A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Detect, cultivate and presumptively identify Mycoplasma hominis and Ureaplasma urealyticum from urogenital specimens, or use it as a transport medium for these organisms.

Detect, cultivate and presumptively identify Mycoplasma hominis and Ureaplasma urealyticum from urogenital specimens with Thermo Scientific™ Mycoplasma / Ureaplasma Broth. It also can be used as a transport medium for these organisms.

The pH of 6.4 is ideal for the growth of Ureaplasma urealyticum. The antibiotic mixture inhibits most gram-negative and gram-positive bacteria as well as yeasts which might be present in the specimens. Mycoplasma species hydrolyse arginine which results in a pH increase because amonia is generated. The color of the broth changes from yellow into orange-red. U. urealyticum is not able to metabolize arginine, but cleaves urea, which results in a pH increase as well. The color of the bouillon turns from yellow into red.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Color Sun yellow, transparent
Content And Storage 2°C to 12°C
Description Mycoplasma / Ureaplasma Broth
Format 90mm tube
Manufacturing Origin Germany
Product Type Microbiology Media
Fill Weight 2mL ± 0.2mL
Packaging Type Flow Wrapped - Single Layer
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.