Learn More
Thermo Scientific™ Mycoplasma / Ureaplasma Agar
Description
Detect, isolate, differentiate and enumerate Mycoplasma and Ureaplasma species mainly from urogenital specimens. Find clarity with an antibiotic mixture in the medium that inhibits most gram-negative and gram-positive bacteria, as well as yeasts, which might be present in patient specimens. On Thermo Scientific™ Mycoplasma / Ureaplasma Agar, the colorless colonies of Mycoplasma hominis form the typical fried egg appearance (growth density-dependent). Colonies of Ureaplasma urealyticum are dark-brown and grow in typical sea urchin morphology.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
| Color | Traffic yellow, transparent |
| Content And Storage | 6°C to 12°C |
| Description | Mycoplasma / Ureaplasma Agar |
| Format | 90mm Monoplate |
| Manufacturing Origin | Germany |
| Product Type | Agar Plate |
| Fill Weight | 13.5g ± 5% |
| Packaging Type | Flow Wrapped - Single Layer |
| Quantity | 10/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.