missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mycoplasma bovis Monoclonal Antibody (MY10-168.5), Invitrogen™
Mouse Monoclonal Antibody
£367.00 - £916.00
Specifications
| Antigen | Mycoplasma bovis |
|---|---|
| Clone | MY10-168.5 |
| Concentration | 1.75 mg/mL |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17810376
|
Invitrogen™
MA5-46110 |
100 μg |
£367.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
11583403
|
Invitrogen™
MA110793 |
500 μg |
£441.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17810346
|
Invitrogen™
MA5-46157 |
1 mg |
£916.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MA1-10793 detects Mycoplasma bovis. MA1-10793 has been successfully used in ELISA applications. The MA1-10793 immunogen is Mycoplasma bovis cells.
This antibody specifically recognize Mycoplasma bovis. It does not cross-react with Mycoplasma genitalium, Mycoplasma californicum, Strep. agalactiae, Staph. aureus or E. coli.Specifications
| Mycoplasma bovis | |
| 1.75 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative; pH 7.4 | |
| Mycoplasma bovis cells. | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles |
| MY10-168.5 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Bacteria | |
| m bovis; M. bovis; mycoplasmen | |
| IgG1 | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title