missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mycoplasma bovis Monoclonal Antibody (MY10-168.5), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA110793
This item is not returnable.
View return policy
Description
MA1-10793 detects Mycoplasma bovis. MA1-10793 has been successfully used in ELISA applications. The MA1-10793 immunogen is Mycoplasma bovis cells.
This antibody specifically recognize Mycoplasma bovis. It does not cross-react with Mycoplasma genitalium, Mycoplasma californicum, Strep. agalactiae, Staph. aureus or E. coli.
Specifications
| Mycoplasma bovis | |
| Monoclonal | |
| 1.75 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Mouse | |
| Protein A | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG1 |
| ELISA | |
| MY10-168.5 | |
| Unconjugated | |
| m bovis; M. bovis; mycoplasmen | |
| Mycoplasma bovis cells. | |
| 500 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction