missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mycobacterium Tuberculosis CFP10 Antibody (KFB16), Novus Biologicals™
Mouse Monoclonal Antibody
£335.00
Specifications
| Antigen | Mycobacterium Tuberculosis CFP10 |
|---|---|
| Clone | KFB16 |
| Dilution | Dot Blot, Sandwich ELISA 1:100-1:2000 |
| Applications | Dot Blot, ELISA |
| Classification | Monoclonal |
Description
Mycobacterium Tuberculosis CFP10 Monoclonal specifically detects Mycobacterium Tuberculosis CFP10 in Bacteria samples. It is validated for ELISA, Dot Blot, Sandwich ELISA.Specifications
| Mycobacterium Tuberculosis CFP10 | |
| Dot Blot, Sandwich ELISA 1:100-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 kDa culture filtrate antigen cfp10, Culture filtrate protein 10, ESAT 6 like protein esxB, esxB, lhp, mtsA10, Secreted antigenic protein MTSA 10, tuberculosis CFP10 | |
| IgG1 | |
| Protein G purified | |
| Mycobacterium tuberculosis CFP10 (KFB16) |
| KFB16 | |
| Dot Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Bacteria | |
| Mycobacterium tuberculosis CFP10 (KFB16) | |
| Primary | |
| Store at 4C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title