missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mycobacterium Tuberculosis CFP10 Antibody (KFB16), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-05466
This item is not returnable.
View return policy
Description
Mycobacterium Tuberculosis CFP10 Monoclonal specifically detects Mycobacterium Tuberculosis CFP10 in Bacteria samples. It is validated for ELISA, Dot Blot, Sandwich ELISA.
Specifications
| Mycobacterium Tuberculosis CFP10 | |
| Monoclonal | |
| Unconjugated | |
| 10 kDa culture filtrate antigen cfp10, Culture filtrate protein 10, ESAT 6 like protein esxB, esxB, lhp, mtsA10, Secreted antigenic protein MTSA 10, tuberculosis CFP10 | |
| Mycobacterium tuberculosis CFP10 (KFB16) | |
| 0.2 mg | |
| Primary | |
| Bacteria | |
| Purified |
| Dot Blot, ELISA | |
| KFB16 | |
| Dot Blot, Sandwich ELISA 1:100-1:2000 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Mycobacterium tuberculosis CFP10 (KFB16) | |
| Store at 4C. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction