missing translation for 'onlineSavingsMsg'
Learn More

Mycobacterium avium subsp. paratuberculosis Antibody (15D10), Novus Biologicals™

Mouse Monoclonal Antibody

£159.00 - £388.00

Specifications

Antigen Mycobacterium avium subsp. paratuberculosis
Clone 15D10
Concentration 1 mg/mL
Dilution ELISA 1:100 - 1:2000, Immunoprecipitation 1:10- 1:500, Competitive ELISA
Applications ELISA, Immunoprecipitation, Competition Assay
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18636826
View Documents
Novus Biologicals
NBP2-54632-0.025mg
0.025 mg
£159.00
0.03mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18650857
View Documents
Novus Biologicals
NBP2-54632
0.1 mg
£388.00
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Mycobacterium avium subsp. paratuberculosis Monoclonal antibody specifically detects Mycobacterium avium subsp. paratuberculosis in Bacteria samples. It is validated for ELISA, Immunoprecipitation, Competitive ELISA
Specifications

Specifications

Mycobacterium avium subsp. paratuberculosis
1 mg/mL
ELISA, Immunoprecipitation, Competition Assay
Unconjugated
RUO
Bacteria
Mycobacterium paratuberculosis
IgM
Thiophilic Chromatography
15D10
ELISA 1:100 - 1:2000, Immunoprecipitation 1:10- 1:500, Competitive ELISA
Monoclonal
Mouse
Virology Bacteria and Parasites
PBS
Ethanol-extracted surface antigens of gamma-irradiated MAP strain B4 cells
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.