missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mycobacterium avium subsp. paratuberculosis Antibody (15D10), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54632
This item is not returnable.
View return policy
Description
Mycobacterium avium subsp. paratuberculosis Monoclonal antibody specifically detects Mycobacterium avium subsp. paratuberculosis in Bacteria samples. It is validated for ELISA, Immunoprecipitation, Competitive ELISA
Specifications
| Mycobacterium avium subsp. paratuberculosis | |
| Monoclonal | |
| 1 mg/mL | |
| ELISA 1:100 - 1:2000, Immunoprecipitation 1:10- 1:500, Competitive ELISA | |
| Mycobacterium paratuberculosis | |
| Ethanol-extracted surface antigens of gamma-irradiated MAP strain B4 cells | |
| 0.1 mg | |
| Virology Bacteria and Parasites | |
| Bacteria | |
| IgM |
| ELISA, Immunoprecipitation, Competition Assay | |
| 15D10 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Thiophilic Chromatography | |
| RUO | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction