missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MVK Monoclonal Antibody (K70044_14G7), Invitrogen™

Mouse Monoclonal Antibody

£416.00 - £734.00

Specifications

Antigen MVK
Clone K70044_14G7
Concentration 1 mg/mL
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30212695
View Documents
Invitrogen™
MA555730
50 μL
£416.00
50µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30203955
View Documents
Invitrogen™
MA556110
200 μL
£734.00
200µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Sequence of this protein is as follows: PGKVILHGEH AVVHGKVALA VSLNLRTFLR LQPHSNGKVD LSLPNIGIKR AWDVARLQSL DTSFLEQGDV TTPTSEQVEK LKEVAGLPDD CAVTERLAVL AFLYLYLSIC RKQRALPSLD IVVWSELPPG AGLGSSAAYS VCLAAALLTV CEEIPNPLKD GDCVNRWTKE DLELINKWAF QGERMIHGNP SGVDNAVSTW GGALRYHQGK ISSLKRSPAL QILLTNTKVP RNTRALVAGV RNRLLKFPEI VAPLLTSIDA ISLECERVLG EMGEAPAPEQ YLVLEELIDM NQHHLNALGV GHASLDQLCQ VTRARGLHSK LTGAGGGGCG ITLLKPGLEQ PEVEATKQAL TSCGFDCLET SIGAPGVSIH SATSLDSRVQ

MVK Monoclonal antibody specifically detects MVK in Human samples. It is validated for Immunoprecipitation, Western Blot
Specifications

Specifications

MVK
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
2310010A05Rik; AI256848; AI414037; KIME; LH receptor mRNA-binding protein; LHR mRNA binding protein; Lrbp; Mevalonate kinase; mevalonate kinase 1; MK; Mvk; MVLK; POROK3
Recombinant human MVK (immunogen range is 11-390).
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Mvk
K70044_14G7
Immunoprecipitation, Western Blot
Unconjugated
Mouse
Human
Q03426
4598
IgG2b
Protein A
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.