Learn More
Invitrogen™ MVK Monoclonal Antibody (K70044_14G7), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA556110
Description
Sequence of this protein is as follows: PGKVILHGEH AVVHGKVALA VSLNLRTFLR LQPHSNGKVD LSLPNIGIKR AWDVARLQSL DTSFLEQGDV TTPTSEQVEK LKEVAGLPDD CAVTERLAVL AFLYLYLSIC RKQRALPSLD IVVWSELPPG AGLGSSAAYS VCLAAALLTV CEEIPNPLKD GDCVNRWTKE DLELINKWAF QGERMIHGNP SGVDNAVSTW GGALRYHQGK ISSLKRSPAL QILLTNTKVP RNTRALVAGV RNRLLKFPEI VAPLLTSIDA ISLECERVLG EMGEAPAPEQ YLVLEELIDM NQHHLNALGV GHASLDQLCQ VTRARGLHSK LTGAGGGGCG ITLLKPGLEQ PEVEATKQAL TSCGFDCLET SIGAPGVSIH SATSLDSRVQ
Mevalonate kinase (MVK) is an early enzyme in isoprenoid and sterol synthesis. Mevalonate kinase catalyzes the ATP-dependent phosphorylation of mevalonic acid to form mevalonate 5-phosphate. Deficiency in MVK activity contributes to mevalonic aciduria and hyperimmunoglobulinemia D/periodic fever syndrome (HIDS).
Specifications
| MVK | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q03426 | |
| Mvk | |
| Recombinant human MVK (immunogen range is 11-390). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunoprecipitation, Western Blot | |
| K70044_14G7 | |
| Unconjugated | |
| Mvk | |
| 2310010A05Rik; AI256848; AI414037; KIME; LH receptor mRNA-binding protein; LHR mRNA binding protein; Lrbp; Mevalonate kinase; mevalonate kinase 1; MK; Mvk; MVLK; POROK3 | |
| Mouse | |
| Protein A | |
| RUO | |
| 4598 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.