Learn More
Honeywell™ Mutex E Light Protective Clothing
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Each
Description
- Antistatic
- Silicone and latex free
- Excellent ergonomics and fitting
- 3-piece hood
- Special crotch design
- Outside flap and dual slider sipper
- Breathability
- 3-layer polypropylene
Specifications
Specifications
| Product Type | Chemical-resistant Coveralls |
| Size | Large |
| Gender | Unisex |
| Material | 3-layer Polypropylene |
| Color | White |
| Protection Class | Protection type: 5, 6 |
| Thickness | 56 gsm |