Learn More
Honeywell™ Mutex 2 Disposable Coveralls with Hood
Product Code. p-7227923
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Each
Pack of 25
Product Code. 11867010
Brand: Honeywell™ 4180002S
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Honeywell™ Mutex 2 Disposable Coveralls offer enhanced protection against tearing and liquid penetration. Constructed from non-woven polypropylene laminated with a microporous film, they are both silicone- and latex-free.
  • Product Type: Coverall
  • Disposable
  • Polypropylene
  • Protection Level: Class 5/6, EC Category 3
  • Liquid and Tear Resistant
  • Non-sterile
  • Latex-Free and Silicone-Free
TRUSTED_SUSTAINABILITY
Specifications
Product Type Single-use Coveralls
Size Small
Gender Unisex
Material Polypropylene
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Hood Type Elastic
Closure Type Dual Slider Zipper
Sterility Non-sterile
Protection Class PPE Directive Category III, Type 5/6
Resistance Liquid, Tears
Certifications/Compliance (Europe) Cat III to Directive 89/686 /EEC, Standard: EN14126 / EN 1073-2 / EN13982-1 / EN13034 Type 6 / EN1149-1
Latex Free Yes
For Use With (Application) Industrial Painting, Pharmaceutical, Microelectronics, Cleanrooms, Biological Hazards, Industrial Cleaning, Asbestos Removal, Chemicals, Petrochemicals
Weight (Metric) 69 g/m2