Description
The multitude of different work steps in the laboratory environment demands a multitude of universally applicable glassware. These glass devices usually have a very simple shape and are characterized by their versatility. Exhibition glasses and museum jars are used to store materials and are often used in exhibitions. Here the objects on display can be preserved and then viewed without touching them.
- The precise grinding of the glass plate and base vessel enables a very tight seal
- Surface plate
Specifications
Specifications
| Dimensions (L x W x H) | 180 x 120 x 60 mm |
| For Use With (Application) | Exhibition of specimen; Storage |
| Material | Borosilicate Glass (3.3) |
| Quantity | 1 Ea. |
| Autoclavable | Autoclavable |