Learn More
MP Biomedicals™ Murashige and Skoog Media
All Murashige and Skoog plant cell culture Media are manufactured from high quality chemicals and reagents.
Brand: MP Biomedicals™ 092623122
Code : NEW
Additional Details : CAS Number : 7757-79-1 Weight : 0.04700kg Transport : UN number : 1479 Chem class : 5.1 Pack group : III
Description
- Each formulation is prepared according to the original specification, except where noted
- Finished lots of media are plant cell tested
Specifications
4°C | |
Murashige and Skoog Basal Medium | |
3.75 to 4.75 |
Powder | |
Solution | |
1 x 10 L |
Safety and Handling
- murashige & skoog basal medium
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Oxidising solid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H272-May intensify fire; oxidiser.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P404-Store in a closed container.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : potassium nitrate, ammonium nitrate, calcium chloride, potassium dihydrogenorthophosphate,zinc sulph