missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Murashige and Skoog Media

Product Code. p-2625456 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 x 1 L
1 x 10 L
1 x 50 L
Product Type:
Murashige and Skoog Basal Medium
Murashige and Skoog Medium
Content And Storage:
0°C to 5°C
4°C
Unit Size:
10 L
50 L
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Product Type Content And Storage Unit Size
17327479 10 x 1 L Murashige and Skoog Basal Medium 0°C to 5°C 10 L
11454925 1 x 10 L Murashige and Skoog Basal Medium 4°C 10 L
11424865 10 x 1 L Murashige and Skoog Medium 4°C 10 L
11434865 1 x 10 L Murashige and Skoog Medium 4°C 10 L
11444865 1 x 50 L Murashige and Skoog Medium 4°C 50 L
17317479 10 x 1 L Murashige and Skoog Basal Medium 4°C 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 17327479 Supplier MP Biomedicals™ Supplier No. 092623220
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

All Murashige and Skoog plant cell culture Media are manufactured from high quality chemicals and reagents.

  • Each formulation is prepared according to the original specification, except where noted
  • Finished lots of media are plant cell tested
TRUSTED_SUSTAINABILITY

Specifications

Form Powder
Product Type Murashige and Skoog Basal Medium
With Additives Gamborg's Vitamins
Quantity 10 x 1 L
Format Solution
pH 3.75 to 4.75
Content And Storage 0°C to 5°C
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Murashige & Skoog Basal Medium with Gamborg's Vitamins
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P235-Keep cool.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P370+P378-In case of fire: Use CO2, powder or waterspray to extinguish.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: potassium nitrate, ammonium nitrate, calcium chloride, boricacid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.