missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Murashige and Skoog Media

All Murashige and Skoog plant cell culture Media are manufactured from high quality chemicals and reagents.

Brand:  MP Biomedicals™ 092610024

Code : NEW

Additional Details : CAS Number : 7757-79-1 Weight : 0.20000kg Transport : UN number : 1477 Chem class : 5.1 Pack group : II


 View more versions of this product

Product Code. 11444865

  • £163.24 / 50 L
Estimated Shipment: 09-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Each formulation is prepared according to the original specification, except where noted
  • Finished lots of media are plant cell tested
Specifications

Specifications

4°C
Murashige and Skoog Medium
Solution
1 x 50 L
Powder
No Sucrose, No Agar, No IAA, No Kinetin
4.80 to 5.65
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Potassium nitrate, ammonium nitrate, calcium chloride, zincsulfate heptahydrate, boric acid

SDS
Documents

Documents

Certificates
Special Offers

Special Offers