Learn More
MP Biomedicals™ Murashige and Skoog Media
All Murashige and Skoog plant cell culture Media are manufactured from high quality chemicals and reagents.
Brand: MP Biomedicals™ 092610022
Code : NEW
Additional Details : Weight : 0.04500kg Transport : UN number : 1477 Chem class : 5.1 Pack group : II
Description
- Each formulation is prepared according to the original specification, except where noted
- Finished lots of media are plant cell tested
Specifications
4°C | |
Murashige and Skoog Medium | |
Solution | |
1 x 10 L |
Powder | |
No Sucrose, No Agar, No IAA, No Kinetin | |
4.80 to 5.65 |
Safety and Handling
- Murashige and Skoog Medium, without Sucrose, Agar, IAA, and Kinetin
Signal Word
- Danger
Hazard Category
- Oxidising solid Category 2
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H272-May intensify fire; oxidiser.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Potassium Nitrate, Ammonium Nitrate, Calcium Chloride, ZincSulfate (Pentahydrate), Boric Acid