missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- 96-well comb block format
- Standard microplate configuration with additional lanes for markers Offset layout of wells allows maximum run length of 1.8cm per well which results in improved separation
- Simplified leak-proof casting dams
- Ultra-compact size minimizes buffer usage and bench space requirements
Specifications
Specifications
| Running Time | 15 to 30 min. |
| Type | Horizontal Gel Electrophoresis System |
| Gel Dimensions | W x L: 10 x 24 cm (3.93 x 9.44 in.) |
| Includes | Electrophoresis unit, UV tray, Comb block with 12 x 8 sample, Combs (1.5mm thick) with one marker lane, casting dams |
| Sample Size | 96 |
| Volume (English) Buffer | 0.18 gal. |
| Volume (Metric) Buffer | 700 mL |
| Width (English) | 18.30 in. |
| Width (Metric) | 46.5 cm |
| Height (English) | 3.14 in. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?