missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ multiSUB™ Midi96 STRETCH Horizontal Gel System

Product Code. 12327287 Shop All Fisherbrand Products
Change view
Click to view available options
Includes:
Electrophoresis unit, UV tray, Comb block with 12 x 8 sample, Combs (1.5mm thick) with one marker lane, casting dams
Electrophoresis unit, UV tray, Comb block with 12 x 8 sample, Combs (1.5mm thick) with two marker lanes, casting dams
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes unitSize
12327287 Electrophoresis unit, UV tray, Comb block with 12 x 8 sample, Combs (1.5mm thick) with two marker lanes, casting dams Each
12337287 Electrophoresis unit, UV tray, Comb block with 12 x 8 sample, Combs (1.5mm thick) with one marker lane, casting dams Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12327287
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Suitable for high throughput electrophoresis of PCR products or samples containing small number of DNA bands.

  • 96-well comb block format
  • Standard microplate configuration with additional lanes for markers
  • Offset layout of wells allows maximum run length of 1.8cm per well which results in improved separation
  • Simplified leak-proof casting dams
  • Ultra-compact size minimizes buffer usage and bench space requirements

Specifications

Running Time 15 to 30 min.
Type Horizontal Gel Electrophoresis System
Gel Dimensions W x L: 10 x 24 cm (3.93 x 9.44 in.)
Includes Electrophoresis unit, UV tray, Comb block with 12 x 8 sample, Combs (1.5mm thick) with two marker lanes, casting dams
Sample Size 96
Volume (English) Buffer 0.18 gal.
Volume (Metric) Buffer 700 mL
Width (English) 18.30 in.
Width (Metric) 46.5 cm
Height (English) 3.14 in.
Height (Metric) 8 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.