missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Multiskan™ FC 96 and 384-well Plate Microplate Photometers

Product Code. 11500695 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
Multiskan FC
Multiskan FC with Incubator
Plate Type:
96 and 384-well Plates
96-well Plate
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Plate Type Unit Size
11500695 Multiskan FC with Incubator 96 and 384-well Plates Each
11590685 Multiskan FC 96-well Plate Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11500695 Supplier Thermo Scientific™ Supplier No. 51119100
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Multiskan FC Microplate Photometer is a reliable instrument for a wide variety of research and routine applications. It reads 96- and 384-well plates and is equipped with shaking and incubation capabilities for temperature critical assays. It can be used as a stand-alone instrument or under PC control with intuitive Thermo Scientific SkanIt software.

Multiskan FC brings together over 40 years experience of the well-known Multiskan product brand, with features for enhanced usability. The Multiskan FC has a 340–850 nm wavelength range, enabling a wide variety of applications from enzyme kinetic studies to Lowry assays. The instrument provides fast and accurate measurements enabling complete 96-well plate reading in less than seven seconds. The instrument is also equipped with linear shaking as standard. A model with an incubator enables incubation temperatures up to 50°C and the capacity to read 384-well plates. SkanIt software supports optimal use of the instrument with a visual workflow, effortless data analysis and exporting capabilities.

Features:

  • Proven performance and reliable day-to-day results through patented optical design and in-built self-diagnostics
  • A broad wavelength range of 340–850 nm for a wide variety of research and routine applications such as ELISA immunoassays, protein quantification, endotoxin, cytotoxicity and proliferation assays, enzyme assays and growth curves
  • Equipped with an eight-position filter wheel with three standard filters, 405 nm, 450 nm and 620 nm, preinstalled
  • A comprehensive range of easy-to install additional filters available for order
  • Fast and accurate measurement of both 96- and 384-well plates for various throughput requirements
  • Shaking and incubation up to 50°C for temperature critical assays
  • Ease of use through the large color screen and a variety of language versions
  • Visual and logical SkanIt software for comprehensive instrument control and data handling
  • Multiskan verification plate available for instrument performance verification
  • Robot compatibility for high-throughput environments
  • Superior usability and logical workflow with Thermo Scientific™ SkanIt™ Software
  • USB port for easy data transfer

Recommended for:

Immunoassays (ELISA), protein assays, endotoxins, cytotoxicity and proliferation assays, enzyme assays, growth curves

TRUSTED_SUSTAINABILITY

Specifications

Product Type Multiskan FC with Incubator
Compatibility Robot
Detector Type Photodetector
Interface On-board or PC Control
Incubator Temperature Ambient +4° to 50°C
Light Source Quartz Halogen Lamp
Measurement Speed 7 sec. for 96-well Plate (Fast Mode), 13 sec. for 96-well Plate (Normal Mode)
Measurement Technology Absorbance
Plate Type 96 and 384-well Plates
Product Line Multiskan
Shaking Linear
Wavelength Selection Filters
Software Type SkanIt Software
Accuracy ± 1% (0 to 3 Abs) or ± 0.003 Abs, which ever is greater at 405 nm
Depth (English) 15.7 in.
Depth (Metric) 40 cm
Description Multiskan FC with incubator
Filters 8-position filter wheel; filters installed: 405 nm, 450 nm, and 620 nm
Height (English) 8.7 in.
Height (Metric) 22 cm
Linearity 96-well plate: 0 to 3 Abs, ±2% at 405 nm, 384-well plate: 0 to 2.5 Abs, ±2% at 405 nm
Precision CV ≤ 0.2% (0.3 to 3 Abs) at 405 nm
Resolution 0.001 Abs
Voltage 100/240 V
Warranty 1 Year
Wavelength Range 340 to 850 nm
Width (English) 11.4 in.
Width (Metric) 29 cm
Frequency 50/60 Hz
Show More Show Less
Can I calibrate the Multiskan FC Microplate Photometer (Fisherbrand accuSkan FC Filter-Based Microplate Photometer)?

The Multiskan FC Microplate Photometer (same instrument as the Fisherbrand accuSkan FC Filter-Based Microplate Photometer) is factory-calibrated and performs self-diagnostics on start-up to ensure consistent results over time. If you need to verify that the instrument is working properly, one of the options below can be utilized:

- Performance can be user-verified using Multiskan Verification Plate (Cat. No. 24072800, for 8 wavelengths or Cat. No. 24072805, for 16 wavelengths).

- The other option for verification is by purchasing our qualification service as an IQ/OQ/PQ or IPV that is required for laboratories that operate within a quality management system, comply with global standards (e.g., GLP, GCP, GMP, ISO 17025, and ISO 15189), and/or follow country-specific regulations (e.g., US FDA 21 CFR Part 11, CLIA, and CAP). Our qualification service provides documented verification that laboratory instruments are installed and operating according to the manufacturer’s performance specifications. More information about this service can be found here.

What other filters are available for the Multiskan FC Microplate Photometer in addition to the three standard filters (405, 450, and 620 nm)?

The Multiskan FC Microplate Photometer offers filters with the following wavelengths for separate purchase. The filter SKU is 142xxx5, where xxx = filter wavelength.
340 nm
375 nm
414 nm
492 nm
520 nm
530 nm
540 nm
550 nm
560 nm
570 nm
595 nm
630 nm
650 nm
690 nm
740 nm
750 nm
820 nm

Is Multiskan FC integrateable with robotics?

Yes, the Multiskan FC is intergrateable with robotics.

My Multiskan FC needs a new lamp. What is the catalog or part number? Can I replace it myself?

Yes, the Halogen lamp 6V/10W is Cat. No. 1410101. You can replace the lamp yourself using instructions in the Multiskan FC User Manual: https://www.thermofisher.com/order/catalog/product/51119100

I think my Multiskan FC isn't working right, but there's no error message. How can I check?

If the results are not as expected, performance can be verified using Multiskan Verification Plate (Cat. Nos. 24072800 (for 8 wavelengths, use with either model) or 24072805 (for 16 wavelengths, use with 51119100)).

Do I need to calibrate my Multiskan FC?

Readers are factory-calibrated and perform self-diagnostics on start-up to ensure consistent results over time. Should an error message appear indicating calibration is needed, contact your local Thermo Fisher Scientific microplate instrument service group at thermofisher.com/contactus.

Can I use uDrop Plate with Multiskan FC?

uDrop Plate is not compatible with Multiskan FC. For UV applications, refer to Multiskan Sky: https://www.thermofisher.com/order/catalog/product/51119500

Can I read my DNA / RNA / protein at 260 / 280 in Multiskan FC?

No, Multiskan FC reads only in the visible range, 340-800 nm. For UV applications, refer to Multiskan Sky: https://www.thermofisher.com/order/catalog/product/51119500

Do I have to use a PC with software to read plates with Multiskan FC?

A computer is not required. The instrument has on-board control that allows for programming and analysis of most assays.

Does the Multiskan FC include software with purchase?

Yes, SkanIt Research Edition is included with purchase; SkanIt Drug Discovery Edition for meeting CFR 21 Part 11 regulations (Cat. No. 5187149) must be purchased separately.

Does the Multiskan FC have a touchscreen?

No, it has a Graphical User Interface with keypad for navigation.

What are the dimensions / weight of Multiskan FC?

The dimensions and weight of the Mjultiskan FC are: ca. 29 cm (W) x 40 cm (D) x 22 cm (H); weight 8.5 kg.

Can I put Multiskan FC in a cold room?

The cold room must meet these operating conditions: +10C to +40C; maximum relative humidity 80% for temperatures up to 31 degrees C decreasing linearly to 50% relative humidity at 40 degrees C, non-condensing; indoor use only.

What are the electrical specifications for Multiskan FC?

The electrical specifications are as follows: 100-240 Vac, 50/60 Hz, nominal.

Is Multiskan FC CE / cSGSus / CSA / NRTL / UL / RoHS certified?

Multiskan FC carries the following certifications: CE, cSGSus, CSA, NRTL, UL, RoHS.

Which plate should I use with Multiskan FC?

We recommend using SBS-standard flat-bottom plates of clear polystyrene that have a total height no taller than 15.3 mm (including any lid). Cat. No. 51119000 can accept 96 well plates; Cat. No. 51119100 can accept either 96 or 384 well plates. U bottom plates are acceptable but not ideal; V bottom plates cannot be used. No specific part number is required as long as it meets the above specifications.

Does the Multiskan FC include shaking?

Yes, both models have linear shaking standard.

Does the Multiskan FC have incubation? What is the range?

The Multiskan FC With Incubator (Cat. No. 51119100) has incubation from ambient +4 to 50 degrees C. The Multiskan FC (Cat. No. 51119000) does not have incubation.

Can the Multiskan FC read 6 / 12 / 24 / 48 well plates?

The Multiskan FC Cat. No. 51119000 can read 96 well plates; the Multiskan FC Cat. No. 51119100 can read 96- and 384-well plates.

Can the Multiskan FC read 384 well plates?

The Multiskan FC With Incubator (Cat. No. 51119100) can read 384-well plates. The Multiskan FC (Cat. No. 51119000) reads only 96-well plates.

What is the linearity of Multiskan FC?

At 405 nm: 0-3 Abs, +/- 2%, 96-well plate; 0-2.5 Abs, +/- 2%, 384-well plate.

How long does it take Multiskan FC to read my plate?

Read speed / measurement speed in Fast mode:
96-well plate = 7 seconds
384-well plate = 13 seconds

What is the half-bandwidth for Multiskan FC filters?

The HBW for a filter will range from 3-9 nm.

How many filters does Multiskan FC hold?

The filter wheel holds up to 8 filters.

What is the part number for additional Multiskan FC filters?

A complete list of available filters with part numbers can be found in the Multiskan FC User Manual which can be found on this page: https://www.thermofisher.com/order/catalog/product/51119100

What filters ship with Multiskan FC?

405, 450 and 650 nm ship with every Multiskan FC. No substitutions are allowed. A complete list of available filters for separate purchase can be found in the Multiskan FC User Manual found on the following page: https://www.thermofisher.com/order/catalog/product/51119100


For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.