missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck MultiScreen™ GFB Filter Plates
£2135.00 - £2285.00
Specifications
Format | Standard |
---|---|
Array | 8 x 12 |
Pore Size | 1 μm |
No. of Wells | 96 |
Sterility | Non-sterile |
Product Code | Brand | For Use With (Application) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Application) | Price | Quantity & Availability | |||||
10094550
|
Merck
MAFCN0B50 |
TCA Precipitation |
£2285.00
Pack of 50 |
Estimated Shipment: 17-09-2024
Log in to see stock. |
|||||
10771981
|
Merck
MAFBN0B50 |
Enzyme Assays with Precipitation |
£2135.00
Pack of 50 |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
Specifications
Standard | |
1 μm | |
Non-sterile | |
0.26 cm2 | |
128 x 85.5 x 14.4 mm | |
85.5 mm |
8 x 12 | |
96 | |
50 to 250 μL | |
50/Pk. | |
128 mm | |
14.4 mm |