missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck MultiScreen™ GFB Filter Plates
Brand: Merck MAFBN0B50
Code : NEW
Additional Details : Weight : 0.01000kg
Specifications
Standard | |
1 μm | |
Non-sterile | |
0.26 cm2 | |
50/Pk. | |
128 mm | |
14.4 mm |
8 x 12 | |
96 | |
50 to 250 μL | |
Enzyme Assays with Precipitation | |
128 x 85.5 x 14.4 mm | |
85.5 mm |