Learn More
WTW™ MultiLine™ Multi 3630 IDS Multi-Parameter Portable Meter
Product Code. 15522026
Change view
Click to view available options
Channels:
3
Unit Size:
Each
Product Code. Channels Unit Size
15522026 3 Each
1 options
Product Code. 15522026 Supplier WTW™ Supplier No. 2FD570
  • ONLINE EXCLUSIVE PRICE
    £2385.00
    £2297.31 / Pack of 1
    Save £87.69
    3% Off
View all promotions
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Solve multiple measuring tasks at once and save time and costs. WTW7™ MultiLine™ Multi 3630 IDS Multi-Parameter Portable Meters feature three universal measuring channels for measuring pH, ORP, dissolved oxygen and/or turbidity simultaneously.
  • Three channels, four parameters
  • Three measuring channels, freely combinable for the same or different parameters
  • For simultaneous multi-parameter measurement without compromise
  • Also use the wireless modules together with the new IDS plug head sensors to be independent from cables and measure conveniently at difficult-to-access locations
  • Allows the operation with the digital multi-parameter probes MPP 910 IDS and MPP 930 IDS
  • Brilliant color graphic display and two USB interfaces
  • The batteries can be charged directly in the device
  • Archiving user administration can also be used alternatively to mark measuring points
  • 10,000 data sets include up to three main parameters, three temperature values as well as the water depth if using the depth probe
TRUSTED_SUSTAINABILITY
Specifications
Outputs Multi 3630 IDS SET F
Backlight Option Yes
Channels 3
Percent Saturation Range (Polarographic DO) Multi 3630 IDS SET KS2
Includes Basic meter unit only with power supply and USB cable
Memory Manual: 500 records ; Automatic: 10,000 records
Model Multi 3630 IDS
Power Supply Wide range power supply, 4 x 1.2 V NiMH battery or via Mini USB-B
Product Type Multi-parameter portable meter