Learn More
WTW™ MultiLine™ Multi 3630 IDS Multi-Parameter Portable Meter Set F
Description
- Three channels, four parameters
- Three measuring channels, freely combinable for the same or different parameters
- For simultaneous multi-parameter measurement without compromise
- Also use the wireless modules together with the new IDS plug head sensors to be independent from cables and measure conveniently at difficult-to-access locations
- Allows the operation with the digital multi-parameter probes MPP 910 IDS and MPP 930 IDS
- Brilliant color graphic display and two USB interfaces
- The batteries can be charged directly in the device
- Archiving user administration can also be used alternatively to mark measuring points
- 10,000 data sets include up to three main parameters, three temperature values as well as the water depth if using the depth probe
Specifications
Specifications
| Backlight Option | Yes |
| Includes | Sensors: SenTix™ 940, FDO™ 925, TetraCon™ 925 |
| Memory | Manual: 500 records ; Automatic: 10,000 records |
| Model | Multi 3630 IDS Set F |
| Power Supply | Wide range power supply, 4 x 1.2 V NiMH battery or via Mini USB-B |
| Product Type | Multi-parameter portable meter |