missing translation for 'onlineSavingsMsg'
Learn More

WTW™ MultiLine™ 3410 Portable Digital Multiparameter

Product Code. 11765184
Change view
Click to view available options
Range (pH):
-2 to 20
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range (pH) unitSize
11765184 -2 to 20 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11765184 Supplier WTW™ Supplier No. 2FD451

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Measure safety without compromise with these portable digital multiparameters. WTW™ Multiline™ 3410 Portable Digital Multiparameters are robust and waterproof (IP 67), ideal for portable field measurements.

  • Digital sensor recognition
  • Trouble-free pH measurements
  • Compliance: IP 67
  • Conductivity - range: 0.0 to 2, 000mS/cm / 0.00 to 19.99μS/cm
  • Connections: USB host and Mini-USB
  • Data logging - capacity:10,000 data sets
  • Data logging - log function: Manual/time controlled
  • Resistivity - range: 0.00 - 20Mω/cm
  • Salinity - range: 0.0 - 70,0ppt
  • TDS - range: 0 - 1, 999mg/L / 0 - 199.9g/L
  • Temperature range (°C): -5.0 +105.0
  • mV - range, ±2, 000 / ±1, 250.0
  • pH - calibration, 1-, 2-, 3-, 4-, 5-point; WTW Technical, DIN/NIST, additionally 20 buffer sets
  • pH - range -2.0.. . +20.0 / -2.00.. . +20.00 / -2.000.. . +20.000
TRUSTED_SUSTAINABILITY

Specifications

Accuracy (Conductivity) ±0.5% full scale
Accuracy (DO) ±0.5% of reading
Accuracy (mV) ±0.2 mV
Accuracy (pH) ±0.004
Accuracy (Resistivity) ±0.5% ±1 digit
Accuracy (Salinity) ±0.5% of reading
Accuracy (TDS) ±0.5% full scale
Battery Type NiMH Rechargeable
Calibration Points Up to 5
Calibration Points (pH) 1, 2, 3, 4, 5-point WTW Technical, DIN/NIST, and 20 additional buffer sets
Certifications/Compliance CE, GLP, ISO 9001:2000
Data Points 10,000
Display Type LCD
IP Rating IP67
Range (Conductivity) 0 to 2000 mS
Range (mV) ±1250, ±2000
Range (pH) -2 to 20
Range (Salinity) 0 to 70 ppt
Range (TDS) 0 to 1999 mg/L
Resolution (pH) 0.1/0.01/0.001
For Use With (Application) Ideal for portable field measurements
Includes Case set with digital IDS pH electrode SenTix™ 940, QSC Kit, short instruction manual, stand, beaker, CD-ROM, driver software for USB, rechargeable batteries, cable, universal power supply and accessories
Memory Automatic, 10,000 data sets
Power Supply Battery charger or 4 x 1.2V NiMH rechargeable batteries
Product Type Portable digital multiparameter
Range 0 to 20mg/L
Resolution (Conductivity) 1μS, 0.1, 1mS
Resolution (mV) 0.1/1
Resolution (Resistivity) 0.01megaohms
Resolution (Salinity) 0.1ppt
Resolution (TDS) 1ppm
Temperature Accuracy ±0.2°C
Temperature Compensation Automatic or manual
Temperature (Metric) Operating -5°C to +105°C
Temperature Resolution 0.1
Warranty 3 Years
Width (Metric) 17.78 cm
Height (Metric) 7.9 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.