Learn More
WTW™ MultiLab™ Pro 40 IDS WL
Product Code. 30929043
Change view
Click to view available options
Channels:
4
Unit Size:
Each
Product Code. Channels Unit Size
30929043 4 Each
1 options
Product Code. 30929043 Supplier WTW™ Supplier No. 1FD78W
  • ONLINE EXCLUSIVE PRICE
    £3680.00
    £3540.97 / Pack of 1
    Save £139.03
    3% Off
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Xylem’s MultiLab™ Pro IDS is a modern high-performance multiparameter device with 4 galvanically isolated measuring channels for pH, ORP, mV, conductivity, oxygen, ISE and turbidity measurement.
The MultiLab™ Pro IDS from Xylem is a modern multi-parameter measuring device with up to four measurement channels.
  • Ease of use
  • LIMS ready
  • GxP compliance
  • Compact and functional design
Highlights:
  • Measures pH, ORP, conductivity, dissolved oxygen, ISE (coming soon) and turbidity
  • Wireless-ready for IDS modules
  • Digital sensor recognition
  • Simultaneous measurement with four independent measuring channels
  • Brilliant touch display
  • Preset and definable buffer sets
  • QSC calibration
  • AutoRead function
  • Export to PDF/A, JSON and CSV
  • Special touchscreen glass can be operated with gloves and resistant to alcohol and disinfectants
Scope of delivery:
  • Device with stand and power supply, IDS WLM-Kit (wireless modules)
TRUSTED_SUSTAINABILITY
Spezifikation
Calibration pH
Certifications/Compliance CE Certified
Channels 4
Display Type Color TFT
IP Rating IP42
Measurement Modes pH, Redox, mV, Conductivity, Oxygen
Range (Conductivity) 0.001 μS/cm to 200 μS/cm
Range (pH) 0.000 to 14.000
Resolution (ISE) depending on Sensor
Resolution (pH) depending on Sensor
Temperature Range (Metric) depending on Sensor
Log Function Type Manual or Interval
Product Type Multiparameter Laboratory Measuring Device
Voltage 115/230 VAC
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.