missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WTW™ MultiLab™ Pro 20 IDS
Click to view available options
Channels:
2
Unit Size:
Each
Description
The MultiLab™ Pro IDS from Xylem is a modern multi-parameter measuring device with up to two measurement channels.
- Ease of use
- LIMS ready
- GxP compliance
- Compact and functional design
.Highlights:
- Measures pH, ORP, conductivity, dissolved oxygen, ISE (coming soon) and turbidity
- Wireless-ready for IDS modules
- Digital sensor recognition
- Simultaneous measurement with up to two independent measuring channels
- GxP-compliant
- Brilliant touch display
- Preset and definable buffer sets
- QSC calibration
- AutoRead function
- LIMS ready
- Export to PDF/A, JSON and CSV
- Special touchscreen glass can be operated with gloves and resistant to alcohol and disinfectants
Scope of delivery:
- Device with stand and power supply
Specifications
Specifications
| Channels | 2 |
| Display Type | Color TFT |
| IP Rating | IP42 |
| Measurement Modes | pH, Redox, mV, Conductivity, Oxygen |
| Range (Conductivity) | 0.001 μS/cm to 200 μS/cm |
| Range (pH) | 0.000 to 14.000 |
| Resolution (ISE) | depending on Sensor |
| Resolution (pH) | depending on Sensor |
| Temperature Range (Metric) | depending on Sensor |
| Log Function Type | Manual or Interval |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction