Learn More
Thermo Scientific™ Mueller-Hinton Agar with 2% Salt
Perform efffortless Antimicrobial Susceptibility Tests (AST) using this ready prepared media plate.
Brand: Thermo Scientific™ PO0799A
Code : 65
Additional Details : Weight : 1.00000kg
Description
Perform effortless Antimicrobial Susceptibility Testing (AST) using Thermo Scientific™ Oxoid™ Mueller Hinton Agar with 2% Salt, a medium designed and manufactured according to the European standard, EUCAST, and the American standard, CLSI. The addition of 2% salt to the medium is recommended when testing staphyococci with oxacillin by either the agar dilution or E test method.1
The major use of Mueller-Hinton Agar with 2% Salt is for Antimicrobial Susceptibility Testing.. The medium is designed and manufactured according to CLSI standards and can be used in internationally recognized standard procedures.
The inclusion of starch in the medium ensures that toxic factors produced during microbial growth will be absorbed, and its presence is often essential to establish growth from very small inocula.
- Specified by the Clinical & Laboratory Standards Institute (CLSI) formerly the National Committee for Clinical Laboratory Standards (NCCLS) 3,4,5 and The European Committee on Antimicrobial Susceptibility Testing (EUCAST) 6.
- Ready-to-use convenience of prepared media.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Straw, clear | |
Mueller-Hinton Agar with 2% Salt | |
United Kingdom | |
26.0g ± 0.5g | |
10/Pk. |
2°C to 10°C | |
90mm Monoplate | |
Agar | |
Flow Wrapped - Single Layer |