Learn More
MUG (4-Methylumbelliferyl-beta-D-glucoronide trihydrate), Fisher Bioreagents™
Product Code. 12801610
Product Code. 12801610 Supplier Fisher Bioreagents Supplier No. 12801610
Only null left
Add to basket
Add to basket
MUG (4-Methylumbelliferyl-beta-D-glucoronide trihydrate) fluorescent substrate for beta-D glucuronidase (GUS) >98% store at 0 to 5°C Fisher BioReagents