missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Mueller Hinton Agar

Product Code. 15852467
Change view
Click to view available options
Quantity:
10/Pk.
4/Pk.
Format:
120mm Squared Plate
90mm Monoplate
Plate
Unit Size:
Pack of 10
Pack of 4
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Format Unit Size
15852467 4/Pk. 120mm Squared Plate Pack of 4
12902588 10/Pk. 90mm Monoplate Pack of 10
15833237 10/Pk. Plate Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15852467 Supplier Thermo Scientific™ Supplier No. PO5403I
  • ONLINE EXCLUSIVE PRICE
    £15.00
    £11.97 / Pack of 4
    Save £3.03
    20% Off
 Restricted Item
In Stock
Ajouter au panier
Ajouter au panier

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Perform effortless Antimicrobial Susceptibility Testing (AST) using this medium which may be used in internationally recognized standard procedures.

Perform effortless Antimicrobial Susceptibility Testing (AST) using Thermo Scientific™ Mueller Hinton Agar, a medium designed and manufactured according to the European standard, EUCAST, and the American standard, CLSI.

The addition of starch to Mueller Hinton Agar ensures that toxic factors found during growth will be absorbed, and its presence is often essential to establish growth from very small inocula. Conforms to EUCAST, CLSI and DIN.

Our products are manufactured and distributed across the world through a vast network of company-owned facilities to ensure you receive the freshest, highest performing medium possible. For fastest delivery We recommend you select the product part number with a manufacturing origin closest to you.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Spécification

Certifications/Compliance According to EUCAST and CLSI
Color Ivory, transparent
Content And Storage 2° to 12°C, in the dark
Description Mueller Hinton Agar
Format 120mm Squared Plate
Manufacturing Origin Germany
Product Type Agar Plate
Fill Weight 60g ± 1.2g
Packaging Type Flow Wrapped - Single Layer
Quantity 4/Pk.
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.