missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Mueller Hinton Agar

Product Code. 15833237
Click to view available options
Quantity:
10/Pk.
4/Pk.
Format:
120mm Squared Plate
90mm Monoplate
Plate
Unit Size:
Pack of 10
Pack of 4
Pack of 5
This item is not returnable. View return policy

Product Code. 15833237

Brand: Thermo Scientific™ PO1191S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Perform effortless Antimicrobial Susceptibility Testing (AST) using this medium which may be used in internationally recognized standard procedures.

Perform effortless Antimicrobial Susceptibility Testing (AST) using Thermo Scientific™ Mueller Hinton Agar, a medium designed and manufactured according to the European standard, EUCAST, and the American standard, CLSI.

The addition of starch to Mueller Hinton Agar ensures that toxic factors found during growth will be absorbed, and its presence is often essential to establish growth from very small inocula. Conforms to EUCAST, CLSI and DIN.

Our products are manufactured and distributed across the world through a vast network of company-owned facilities to ensure you receive the freshest, highest performing medium possible. For fastest delivery We recommend you select the product part number with a manufacturing origin closest to you.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 10°C
Format Plate
Product Type Agar
Quantity 10/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.