missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Mueller Hinton Agar with Horse Blood

Product Code. p-7000007
Click to view available options
Quantity:
10/Pk.
Format:
90mm Monoplate
Media Type:
MHA with Horse Blood
Manufacturing Origin:
United Kingdom
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 15574989

Brand: Thermo Scientific™ PB1229A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Perform effortless Antimicrobial Susceptibility Testing (AST) using this medium for fastidious organisms according to EUCAST.

Perform effortless Antimicrobial Susceptibility Testing (AST) using Thermo Scientific™ Mueller Hinton Agar with Horse Blood, a medium designed and manufactured according to the European standard, EUCAST.

The addition of horse blood and NAD enable the growth of fastidious organisms and the formulation ensures that toxic factors will be absorbed to establish better growth from very small inocula. Conforms to EUCAST.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need, and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance According to EUCAST
Color Red, opaque
Content And Storage 2°C to 10°C
Format 90mm Monoplate
Manufacturing Origin United Kingdom
Media Type MHA with Horse Blood
Product Type Agar
Fill Weight 26g ± 0.6g
Packaging Type Flow Wrapped - Single Layer
Quantity 10/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.