missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Mueller Hinton Agar with Horse Blood
Description
Perform effortless Antimicrobial Susceptibility Testing (AST) using Thermo Scientific™ Mueller Hinton Agar with Horse Blood, a medium designed and manufactured according to the European standard, EUCAST.
The addition of horse blood and NAD enable the growth of fastidious organisms and the formulation ensures that toxic factors will be absorbed to establish better growth from very small inocula. Conforms to EUCAST.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need, and the quality results your laboratory depends on.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need, and the quality results your laboratory depends on.
Specifications
Specifications
| Certifications/Compliance | According to EUCAST |
| Color | Red, opaque |
| Content And Storage | 2°C to 10°C |
| Format | 90mm Monoplate |
| Manufacturing Origin | United Kingdom |
| Media Type | MHA with Horse Blood |
| Product Type | Agar Plate |
| Fill Weight | 26g ± 0.6g |
| Packaging Type | Flow Wrapped - Single Layer |
| Quantity | 10/Pk. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?