missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTUS1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£139.00 - £299.00
Specifications
| Antigen | MTUS1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18316965
|
Novus Biologicals
NBP3-15462-20UL |
20 μg |
£139.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18312087
|
Novus Biologicals
NBP3-15462-100UL |
100 μg |
£299.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MTUS1 Polyclonal antibody specifically detects MTUS1 in Human samples. It is validated for Western BlotSpecifications
| MTUS1 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Angiotensin-II type 2 receptor-interacting protein, AT2 receptor-binding protein, AT2R binding protein, ATBPATIP1, ATIP, DKFZp586D1519, erythroid differentiation-related, FLJ14295, GK1, KIAA1288DKFZp686F20243, microtubule associated tumor suppressor 1, microtubule-associated tumor suppressor 1, Mitochondrial tumor suppressor 1, mitochondrial tumor suppressor gene 1, MP44, MTSG1AT2 receptor-interacting protein, transcription factor MTSG1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 207-436 of human MTUS1 (NP_065800.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Breast Cancer | |
| PBS, 50% glycerol, pH7.3 | |
| 57509 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title