missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTUS1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-15462-20UL
This item is not returnable.
View return policy
Description
MTUS1 Polyclonal antibody specifically detects MTUS1 in Human samples. It is validated for Western Blot
Specifications
| MTUS1 | |
| Polyclonal | |
| PBS, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| Angiotensin-II type 2 receptor-interacting protein, AT2 receptor-binding protein, AT2R binding protein, ATBPATIP1, ATIP, DKFZp586D1519, erythroid differentiation-related, FLJ14295, GK1, KIAA1288DKFZp686F20243, microtubule associated tumor suppressor 1, microtubule-associated tumor suppressor 1, Mitochondrial tumor suppressor 1, mitochondrial tumor suppressor gene 1, MP44, MTSG1AT2 receptor-interacting protein, transcription factor MTSG1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 207-436 of human MTUS1 (NP_065800.1). | |
| 20 μg | |
| Breast Cancer | |
| 57509 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction