missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ MTT Reagent

Product Code. 30000924
Change view
Click to view available options
Quantity:
25 mL
Unit Size:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30000924 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30000924 Supplier R&D Systems™ Supplier No. 48902501

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

R&D Systems™ MTT Reagent yields low background absorbance values in the absence of cells and is stable when stored at 4°C.

  • Contains less than 1% (w/v) MTT (3-(4,5-dimethylthiazolyl-2)-2, 5-diphenyl-tetrazolium bromide (CAS # 298-93-1).
  • MTT is toxic and may cause heritable genetic defects. In case of contact, immediately flush eyes or skin with copious amounts of water. If swallowed, wash out mouth with water provided person is conscious
  • For use with  MTT Cell Proliferation Assays
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Quantity 25 mL
Sample Volume 25 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.