missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTMR15 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17600-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
MTMRF encodes a member of the myotubularin-related class 1 cysteine-based protein tyrosine phosphatases. The encoded protein may be catalytically inactive.Specifications
MTMR15 | |
Polyclonal | |
Unconjugated | |
FAN1 | |
FAN1, Fanconi associated nuclease 1, KIAA1018, MTMR15 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22909 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9Y2M0 | |
FAN1 | |
MTMR15 Fusion Protein Ag11651 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |