missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTMR15 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£130.00 - £339.00
Specifications
Antigen | MTMR15 |
---|---|
Concentration | 0.21 mg/mL |
Applications | Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16877534
|
Proteintech
17600-1-AP-20UL |
20 μL |
£130.00
20µL |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16867534
|
Proteintech
17600-1-AP-150UL |
150 μL |
£339.00
150µL |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
MTMRF encodes a member of the myotubularin-related class 1 cysteine-based protein tyrosine phosphatases. The encoded protein may be catalytically inactive.Specifications
MTMR15 | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q9Y2M0 | |
22909 | |
MTMR15 Fusion Protein Ag11651 | |
Primary | |
-20°C | |
FAN1 |
0.21 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
FAN1, Fanconi associated nuclease 1, KIAA1018, MTMR15 | |
FAN1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |