missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTMR15 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£369.00
Specifications
| Antigen | MTMR15 |
|---|---|
| Dilution | Western Blot 1.0 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
MTMR15 Polyclonal specifically detects MTMR15 in Human samples. It is validated for Western Blot.Specifications
| MTMR15 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| DKFZp451H236, EC 3.1.21.-, EC 3.1.4.1, FANCD2/FANCI-associated nuclease 1DKFZp686K16147, fanconi anemia associated nuclease 1, fanconi-associated nuclease 1, KIAA1018coiled-coil domain-containing protein MTMR15, MTMR15, myotubularin related protein 15, Myotubularin-related protein 15 | |
| The immunogen is a synthetic peptide directed towards the C terminal region of human MTMR15 (NP_001139568.1). Peptide sequence QVGLINSNVSMVDLTSVTLEDVTPKKSPPPKTNLTPGQSDSAKREVKQKI | |
| Affinity purified |
| Western Blot 1.0 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS buffer, 2% sucrose | |
| 22909 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title