missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTMR12 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£169.00 - £255.00
Specifications
| Antigen | MTMR12 |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18660922
|
Novus Biologicals
NBP2-97310-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18673234
|
Novus Biologicals
NBP2-97310-50ul |
50 μg |
£169.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MTMR12 Polyclonal antibody specifically detects MTMR12 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| MTMR12 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Protein Phosphatase | |
| PBS (pH 7.0) | |
| 54545 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| 3-PAP3PAP, 3-phosphatase adapter subunit, KIAA1682FLJ20476, myotubularin related protein 12, myotubularin-related protein 12,3-phosphatase adapter protein, Phosphatidylinositol-3 phosphate 3-phosphatase adapter subunit, phosphatidylinositol-3 phosphate 3-phosphatase adaptor subunit, phosphatidylinositol-3-phosphate associated protein, PIP3AP | |
| Produced in rabbits immunized with E. coli-derived Human MTMR12 fragment. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title