missing translation for 'onlineSavingsMsg'
Learn More

MTMR12 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-97310-50ul

 View more versions of this product

Product Code. 18673234

  • £169.00 / 50µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MTMR12 Polyclonal antibody specifically detects MTMR12 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MTMR12
Polyclonal
Immunohistochemistry-Paraffin 1:50-1:200
3-PAP3PAP, 3-phosphatase adapter subunit, KIAA1682FLJ20476, myotubularin related protein 12, myotubularin-related protein 12,3-phosphatase adapter protein, Phosphatidylinositol-3 phosphate 3-phosphatase adapter subunit, phosphatidylinositol-3 phosphate 3-phosphatase adaptor subunit, phosphatidylinositol-3-phosphate associated protein, PIP3AP
Produced in rabbits immunized with E. coli-derived Human MTMR12 fragment.
50 μg
Protein Phosphatase
54545
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Immunohistochemistry (Paraffin)
Unconjugated
PBS (pH 7.0)
Rabbit
Antigen and protein A Affinity-purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.