missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTMR12 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-55211
This item is not returnable.
View return policy
Description
MTMR12 Polyclonal specifically detects MTMR12 in Human samples. It is validated for Western Blot.
Specifications
| MTMR12 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| 3-PAP3PAP, 3-phosphatase adapter subunit, KIAA1682FLJ20476, myotubularin related protein 12, myotubularin-related protein 12,3-phosphatase adapter protein, Phosphatidylinositol-3 phosphate 3-phosphatase adapter subunit, phosphatidylinositol-3 phosphate 3-phosphatase adaptor subunit, phosphatidylinositol-3-phosphate associated protein, PIP3AP | |
| Rabbit | |
| 86 kDa | |
| 100 μL | |
| Protein Phosphatase | |
| 54545 | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9C0I1 | |
| MTMR12 | |
| Synthetic peptides corresponding to MTMR12(myotubularin related protein 12) The peptide sequence was selected from the middle region of MTMR12. Peptide sequence PLYVEKPKLDKGQRKGMRFKHQRQLSLPLTQSKSSPKRGFFREETDHLIK. | |
| Affinity purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction