missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTG1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | MTG1 |
|---|---|
| Concentration | 0.17 mg/mL |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16862114
|
Proteintech
13742-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16852114
|
Proteintech
13742-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Mitochondrial GTPase. May be involved in assembly of the large ribosomal subunit.Specifications
| MTG1 | |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat, Human | |
| Q8R2R6, Q9BT17 | |
| 100910751, 212508, 92170 | |
| MTG1 Fusion Protein Ag4681 | |
| Primary | |
| -20°C | |
| MTG1 |
| 0.17 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| GTP, GTP binding protein 7, GTPBP7, Mitochondrial GTPase 1, MTG1, RP11 108K14.2 | |
| MTG1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title