missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTG1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13742-1-AP-150UL
This item is not returnable.
View return policy
Description
Mitochondrial GTPase. May be involved in assembly of the large ribosomal subunit.Specifications
| MTG1 | |
| Polyclonal | |
| Unconjugated | |
| MTG1 | |
| GTP, GTP binding protein 7, GTPBP7, Mitochondrial GTPase 1, MTG1, RP11 108K14.2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100910751, 212508, 92170 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8R2R6, Q9BT17 | |
| MTG1 | |
| MTG1 Fusion Protein Ag4681 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction