missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTF2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16208-1-AP-20UL
This item is not returnable.
View return policy
Description
Polycomb group (PcG) that specifically binds histone H3 trimethylated at ′Lys-36′ (H3K36me3) and recruits the PRC2 complex. Acts by binding to H3K36me3, a mark for transcriptional activation, and recruiting the PRC2 complex, leading to enhance PRC2 H3K27me3 methylation activity. Regulates the transcriptional networks during embryonic stem cell self-renewal and differentiation. Promotes recruitment of the PRC2 complex to the inactive X chromosome in differentiating XX ES cells and PRC2 recruitment to target genes in undifferentiated ES cells. Required to repress Hox genes by enhancing H3K27me3 methylation of the PRC2 complex. In some conditions may act as an inhibitor of PRC2 activity: able to activate the CDKN2A gene and promote cellular senescence by suppressing the catalytic activity of the PRC2 complex locally. Binds to the metal-regulating-element (MRE) of MT1A gene promoterSpecifications
| MTF2 | |
| Polyclonal | |
| Unconjugated | |
| MTF2 | |
| hPCl2, M96, MTF2, PCL2, Polycomb like protein 2, RP5 976O13.1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 17765, 22823, 360905 | |
| -20°C | |
| Liquid |
| Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q02395, Q9Y483 | |
| Mtf2 | |
| MTF2 Fusion Protein Ag9182 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction