missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTAP (Tumor Suppressor Marker) Mouse, Clone: MTAP/1813, enQuireBio™
Mouse Monoclonal Antibody
£181.83 - £395.00
Specifications
Antigen | MTAP (Tumor Suppressor Marker) |
---|---|
Clone | MTAP/1813 |
Concentration | 200μg/mL |
Applications | ELISA, Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15960498
|
EnquireBio
4507-MSM1-P1 |
100 μg |
£395.00
100µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15950498
|
EnquireBio
4507-MSM1-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
MTAP (Tumor Suppressor Marker) | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant human MTAP protein fragment (aa97-196) | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
31kDa |
MTAP/1813 | |
ELISA, Western Blot, Immunohistochemistry | |
Unconjugated | |
RUO | |
4507 | |
IgG2b κ | |
Protein A/G purified from Bioreactor concentrate. | |
Cytoplasmic | |
HepG2, A431, HeLa or MCF-7 cells. Kidney |