missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTAP (Tumor Suppressor Marker) Mouse, Clone: MTAP/1813, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 4507-MSM1-P0
Additional Details : Weight : 0.01000kg
Specifications
MTAP (Tumor Suppressor Marker) | |
Cytoplasmic | |
MTAP/1813 | |
Unconjugated | |
Mouse | |
31kDa | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
4507 | |
IgG2b κ |
ELISA, Western Blot, Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant human MTAP protein fragment (aa97-196) | |
HepG2, A431, HeLa or MCF-7 cells. Kidney | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |