missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSPR/Ron Antibody (CL0463), Novus Biologicals™
Mouse Monoclonal Antibody
£292.00 - £435.00
Specifications
| Antigen | MSPR/Ron |
|---|---|
| Clone | CL0463 |
| Dilution | Western Blot 1 ug/ml |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18498861
|
Novus Biologicals
NBP2-30924-25ul |
25 μL |
£292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18131114
|
Novus Biologicals
NBP2-30924 |
0.1 mL |
£435.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MSPR/Ron Monoclonal specifically detects MSPR/Ron in Human samples. It is validated for Western Blot.Specifications
| MSPR/Ron | |
| Western Blot 1 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD136, CD136 antigen, CDw136c-met-related tyrosine kinase, EC 2.7.10, EC 2.7.10.1, macrophage stimulating 1 receptor (c-met-related tyrosine kinase), MSP receptor, p185-Ron, Protein-tyrosine kinase 8, PTK8, PTK8 protein tyrosine kinase 8, RONmacrophage-stimulating protein receptor, soluble RON variant 1, soluble RON variant 2, soluble RON variant 3, soluble RON variant 4 | |
| MST1R | |
| IgG1 | |
| Protein A purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| CL0463 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cell Cycle and Replication, GPCR, Immune System Diseases, Immunology, Protein Kinase | |
| Q04912 | |
| 4486 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: EDPVVLSISPNCGYINSHITICGQHLTSAWHLVLSFHDGLRAVESRCERQLPEQQLCRLPEYVVRDPQGWVAGNLSARGDGAAGFTLPGFRFLPPPHPPSANLVPLKP | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title