missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSPR/Ron Antibody (CL0463), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-30924
This item is not returnable.
View return policy
Description
MSPR/Ron Monoclonal specifically detects MSPR/Ron in Human samples. It is validated for Western Blot.
Specifications
| MSPR/Ron | |
| Monoclonal | |
| Unconjugated | |
| Q04912 | |
| MST1R | |
| This antibody was developed against a recombinant protein corresponding to amino acids: EDPVVLSISPNCGYINSHITICGQHLTSAWHLVLSFHDGLRAVESRCERQLPEQQLCRLPEYVVRDPQGWVAGNLSARGDGAAGFTLPGFRFLPPPHPPSANLVPLKP | |
| 0.1 mL | |
| Apoptosis, Cell Cycle and Replication, GPCR, Immune System Diseases, Immunology, Protein Kinase | |
| 4486 | |
| Human | |
| Purified |
| Western Blot | |
| CL0463 | |
| Western Blot 1 ug/ml | |
| CD136, CD136 antigen, CDw136c-met-related tyrosine kinase, EC 2.7.10, EC 2.7.10.1, macrophage stimulating 1 receptor (c-met-related tyrosine kinase), MSP receptor, p185-Ron, Protein-tyrosine kinase 8, PTK8, PTK8 protein tyrosine kinase 8, RONmacrophage-stimulating protein receptor, soluble RON variant 1, soluble RON variant 2, soluble RON variant 3, soluble RON variant 4 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction