missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSK1/RPS6KA5, p Ser376 Antibody (2F3), Novus Biologicals™
Rabbit Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | MSK1/RPS6KA5 |
|---|---|
| Clone | 2F3 |
| Dilution | ELISA, Immunohistochemistry 1:50-1:200 |
| Applications | ELISA, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30030347
|
Novus Biologicals
NBP3-26393-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30025898
|
Novus Biologicals
NBP3-26393-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MSK1/RPS6KA5 Monoclonal antibody specifically detects MSK1/RPS6KA5 in Human samples. It is validated for ELISA, ImmunohistochemistrySpecifications
| MSK1/RPS6KA5 | |
| ELISA, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 2.7.11, MGC1911,90 kDa ribosomal protein S6 kinase 5, MSK1EC 2.7.11.1, MSPK1, Nuclear mitogen- and stress-activated protein kinase 1, ribosomal protein S6 kinase alpha-5, ribosomal protein S6 kinase, 90kD, polypeptide 5, ribosomal protein S6 kinase, 90kDa, polypeptide 5, RLPK, RSKL, RSK-like protein kinase, S6K-alpha-5 | |
| A synthesized peptide derived from Human MSK1/RPS6KA5 [p Ser376] [UniProt O75582] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 2F3 | |
| ELISA, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Phospho Specific, Protein Kinase, Signal Transduction | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 9252 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title